Order Entry
United States
ContactUsLinkComponent
Drum Pump, 8, 10 Or 12 Oz Per Stroke, Ryton 316SS Rod And PTFE
Drum Pump, 8, 10 Or 12 Oz Per Stroke, Ryton 316SS Rod And PTFE
Catalog # MFLX06511-80
Supplier:  Action Pump
undefined
Drum Pump, 8, 10 Or 12 Oz Per Stroke, Ryton 316SS Rod And PTFE
Catalog # MFLX06511-80
Supplier:  Action Pump
Supplier Number:  R490ST

About this item

Economical pumps for use in cabinets or tight work spaces.

  • Fit 15, 30, and 55-gallon drums with 2" IPS bung openings
  • Designed for use in industrial, agricultural, labs, and automotive industries

Choose from center-level pump and lever-action pump model options.

Center-Lever Pump is self-priming and features an anti-drip spout to eliminate wasted fluid. This pump is ideal for oils, detergents, and other compatible industrial fluids.

Lever-Action Pumps feature a curved discharge spout for convenient hose extension. Choose from three models depending on your chemical needs. Model 06511-50 is ideal for water, alcohol, gasoline, diesel, kerosene, and lubricants. Model 06511-60 handles the same fluids, plus water-based chemicals, acids, and alkalis. Model 06511-80 handles the same fluids, plus strong acids and strong alkalis.

Lever-Action Pumps feature a curved discharge spout or convenient hose extension. Choose from three models depending on your chemical needs. Model 06511-50 is ideal for water, alcohol, gasoline, diesel, kerosene, and lubricants. Model 06511-60 handles the same fluids, plus water-based chemicals, acid, and alkaline. Model 06511-80 handles the same fluids, plus strong acids and strong alkalis.

  • Flow rate:
    16 strokes/gal.
  • Description:
    Drum Pump, 8, 10 Or 12 Oz Per Stroke, Ryton 316SS Rod And PTFE
  • Environmentally Preferable:
  • Wetted Materials:
    Ryton® body with Stainless Steel rod, PTFE seals
  • Tube length, inlet:
    36.5"
  • Warranty:
    3 Months
  • Cat. No.:
    MFLX06511-80
Frequently Bought Together