Universal Tapered Tubing Adapter; 1/Each
Catalog # MFLX06432-00
Supplier: VWR International
About this item
Forms a continuous standard taper inside and out, from top to bottom.
- Cut into various adapters, such as individual bushings, stirrer bearings, thermometer and gas inlet tube supports, drip-tips, funnels, trap connectors for tubing, etc.
Polypropylene cone is 27-cm (10-5/8") long; cut to fit your needs. Tapers from 30- to 3-mm ID (35- to 8-mm OD). Withstands temperatures to 250ºF (121ºC). Bel-Art.
- Description:Universal Tapered Tubing Adapter
- Material:Acetal
- Environmentally Preferable:
- Cat. No.:MFLX06432-00
Frequently Bought Together