Hedland DC Switch for spring-loaded in-line flowmeters, 1 A max, 10 to 30 VDC
Catalog # MFLX03231-85
Supplier: BADGER METER INC
About this item
Make the most of your Hedland flowmeters with these replacement parts and other accessories.
- Optimize performance of your flowmeter with these accessories
Replacement switches and cables are suitable for use with Hedland spring-loaded in-line flowmeters or turbine flowmeters.
- Description:DC Switch for spring-loaded in-line flowmeters, 1 A max, 10 to 30 VDC
- Environmentally Preferable:
- Warranty:1 year
- Cat. No.:MFLX03231-85
Frequently Bought Together