Order Entry
United States
ContactUsLinkComponent
Hedland AC Switch for spring-loaded in-line flowmeters, 0.5 A max, 115 VAC
Hedland AC Switch for spring-loaded in-line flowmeters, 0.5 A max, 115 VAC
Catalog # MFLX03231-80
Supplier:  BADGER METER INC
undefined
Hedland AC Switch for spring-loaded in-line flowmeters, 0.5 A max, 115 VAC
Catalog # MFLX03231-80
Supplier:  BADGER METER INC
Supplier Number:  H526-003
  • Warranty:
  • 1 year
  • 1 year
  • Currently Viewing

Specifications

About this item

Make the most of your Hedland flowmeters with these replacement parts and other accessories.

  • Optimize performance of your flowmeter with these accessories

Replacement switches and cables are suitable for use with Hedland spring-loaded in-line flowmeters or turbine flowmeters.

  • Description:
    AC Switch for spring-loaded in-line flowmeters. 0.5 A max. 115 VAC
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Cat. No.:
    MFLX03231-80
Frequently Bought Together