Masterflex® Flowmeter Acrylic Tripod Base for 03218-50 and 03218-52
Catalog # MFLX03218-58
Supplier: AALBORG INSTRUMENTS & CONTROL
About this item
Easily mount flowmeters on tripod for easy viewing.
- Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
- Description:Tripod Base for 03218-50 and 03218-52
- Environmentally Preferable:
- Model:03218-58
- Width:1755"
- Warranty:1 year
- Cat. No.:MFLX03218-58
- Housing material:Acrylic
Frequently Bought Together