SEPARATION MODULE CARTRID 2X13 2-40 KDA
Catalog # MSPP-SM-W009
Supplier: PROTEIN SIMPLE MS
SEPARATION MODULE CARTRID 2X13 2-40 KDA
Catalog # MSPP-SM-W009
Supplier: PROTEIN SIMPLE MS
Supplier Number: SM-W009
Frequently Bought Together
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® UHP Multichannel Pipettes
Supplier: VWR International
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Sodium chloride 5 M, sterile-filtered
Supplier: Teknova
Costar® Low Binding Microcentrifuge Tubes, Polypropylene, Corning
Supplier: Corning
Inserts for High Recovery Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Gas Clean Purification System, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CARTRIDGE FLUORESCENCE MODULE 12-230 KDA
Supplier: 0000014126
Anti-Rabbit Detection Module, Protein Simple
Supplier: PROTEIN SIMPLE MS
Plasmid+® Media, Thomson Instrument Company
Supplier: THOMSON INSTRUMENT CO
RESIN STREP-TACTIN XT 4FLOW HIGH 20ML
Supplier: IBA MS
DETECTION MODULE NO SECONDARY
Supplier: 0000014126
Sodium chloride 0.9% in water normal saline solution
Supplier: G-Biosciences
RESIN STREPTACTIN XT 4FLOW 2-8DEG C 50ML
Supplier: IBA MS
PI MARKER ICE280/ICE3 4.65 200UL
Supplier: 0000014126
CARTRIDGE PREP C4-300A 15X21.2MM/ID
Supplier: 0000013469
COLUMN LC HYDRO-RP 80A 4UM 150X3MM
Supplier: 0000013469