HUMAN LDL R TAG FREE (LDR-H5215) 100uG
Catalog # 104015-096
Supplier: ACRO Biosystems
HUMAN LDL R TAG FREE (LDR-H5215) 100uG
Catalog # 104015-096
Supplier: ACRO Biosystems
Supplier Number: LDRH5215100UG
Frequently Bought Together
Potassium chloride 3.0 M in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics