FILTER ASSEMBLY FLOW PATH FOR P1
Catalog # 76522-072
Supplier: HITACHI HIGH-TECH AMERICA, INC.
FILTER ASSEMBLY FLOW PATH FOR P1
Catalog # 76522-072
Supplier: HITACHI HIGH-TECH AMERICA, INC.
Supplier Number: 852-1537
Frequently Bought Together
Rotors for ROTOFIX 32A Centrifuge, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
Charts and Pens for 6" Dickson® Chart Recorders, Graphic Controls
Supplier: Graphic Controls
RACK 1X100ML D46MM RB PK2
Supplier: Ohaus
Thermometer, Organic Liquid-Filled, Ace Glass
Supplier: Ace Glass
Eppendorf® 5810 R Refrigerated Benchtop Centrifuge
Supplier: Eppendorf
Eppendorf® 5430 Microcentrifuges
Supplier: Eppendorf
Project Timer, Sper Scientific
Supplier: Sper Scientific
Accessories for Eppendorf®, Adapter, Multipurpose Centrifuges 5804, 5804R
Supplier: Eppendorf
SHIELD OX ANTI TRIP HERCULES
Supplier: LINEMASTER SWITCH CORP MS
Food Safety Thermometer with IR, Certified, Sper Scientific
Supplier: Sper Scientific
Vernier® Relative Humidity Sensor
Supplier: Vernier Software & Technology
Benchtop Centrifuges and Packages, Multifuge X1 and X1R Pro, Thermo Scientific
Supplier: Thermo Fisher Scientific
SEAL L-2200 INJECTION VALVE F/AUTOSAMPLR
Supplier: HITACHI HIGH-TECH AMERICA, INC.
Rotors for Centrifuge 5430/5430 R
Supplier: Eppendorf
HIGH BLOCK COVER
Supplier: G-Biosciences
AMINO ACID ANALYZER PF1 BUFFER
Supplier: HITACHI HIGH-TECH AMERICA, INC.
Accessories for Allegra® V-15R Benchtop Centrifuges, Beckman Coulter
Supplier: Beckman Coulter
AllSpin™ JS-5.3 Swinging Bucket Rotors, Beckman Coulter®
Supplier: Beckman Coulter
ADAPTER 1X50ML D29MM RB V1 PK2
Supplier: Ohaus