MAB SARS-COV-2 SPIKE RBD AM122 100UG
Catalog # 104014-514
Supplier: ACROBIOSYSTEMS
MAB SARS-COV-2 SPIKE RBD AM122 100UG
Catalog # 104014-514
Supplier: ACROBIOSYSTEMS
Supplier Number: SPDM201100UG
Frequently Bought Together
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
VWR® Weighing Paper
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
CiDehol® ST Sterile 70% IPA with WFI, Decon Labs
Supplier: Decon Labs
VWR® Disposable Petri Dishes
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
SARS-CoV-2 Recombinant S protein RBD (from HEK293 cells)
Supplier: ACROBIOSYSTEMS