BUFFER BORG DECLOAKER RTU 1GALLON
Catalog # MSPP-BD1000G1
Supplier: BIOCARE MEDICAL MS
BUFFER BORG DECLOAKER RTU 1GALLON
Catalog # MSPP-BD1000G1
Supplier: BIOCARE MEDICAL MS
Supplier Number: BD1000G1
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Histology Cassettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Micro Cover Glasses, Rectangular
Supplier: VWR International
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Paraffin Tissue Microarrays, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
HEMATOXYLIN NOVOCASTRA 25ML COUNTERSTAIN
Supplier: Leica Biosystems MS
RNA ScreenTape Analysis
Supplier: Agilent Technologies
Enviro-Safe® Thermometer, Organic Black Liquid Filled
Supplier: Ace Glass
KIT DETECTION GOAT IGG POLYMER HRP 50ML
Supplier: Vector Laboratories
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes, Epredia
Supplier: Epredia
Decloaking Chamber™ Antigen Retrieval Chamber
Supplier: BIOCARE MEDICAL MS
REAGENT KIT BETAZOID DAB CHROMOGEN 100ML
Supplier: 0000019526