Order Entry
United States
Orders LinkContactUsLinkComponent
GenScript® 4X LDS Sample Buffer, GenScript

GenScript® 4X LDS Sample Buffer, GenScript

4X LDS sample buffer 250 ml, 250 ml
Catalog # 76510-612
Supplier:   Genscript
Supplier #:  M00676-250
GenScript® 4X LDS Sample Buffer, GenScript
Catalog # 76510-612
Supplier:  Genscript
CAS Number:  
GenScript® 4X LDS Sample Buffer, GenScript
GenScript® 4X LDS Sample Buffer, GenScript

4X LDS sample buffer 250 ml, 250 ml

CAS Number:  
Price
$154.43
Log in for availability
Each (250ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Documents

Specifications

Specifications

  • Description:
    4X LDS sample buffer 250 ml
  • Size:
    250 ml

Specifications