Order Entry
United States
Orders LinkContactUsLinkComponent
GenScript® 4X LDS Sample Buffer, GenScript

GenScript® 4X LDS Sample Buffer, GenScript

4X LDS sample buffer 10 ml, 10 ml
Catalog # 76510-610
Supplier:   Genscript
Supplier #:  M00676-10
GenScript® 4X LDS Sample Buffer, GenScript
Catalog # 76510-610
Supplier:  Genscript
CAS Number:  
GenScript® 4X LDS Sample Buffer, GenScript
GenScript® 4X LDS Sample Buffer, GenScript

4X LDS sample buffer 10 ml, 10 ml

CAS Number:  
Price
$22.36
Log in for availability
Each (10ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Documents

Specifications

Specifications

  • Description:
    4X LDS sample buffer 10 ml
  • Size:
    10 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.