CrossLab Dissolution Paddles, Agilent Technologies
Paddle, lower interchangeable Spin on/off, electropolished stainless steel, 1 L, 21", for 705-DS; 708-DS; 709-DS; VK7000
Catalog # AG14-3603
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
: 14-3603
CrossLab Dissolution Paddles, Agilent Technologies
Catalog # AG14-3603
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Jump to:
Product Details & Documents
Available in stainless steel, PTFE-coated, or PEEK in a range of sizes.
- All paddles are serialized and designed to meet USP specifications for Apparatus 2
- Select from electropolished stainless steel, PTFE-coated, or the biocompatible, chemically inert PEEK paddles in a range of sizes
- Additionally, mini and mega paddles for small- and large-volume dissolution testing are available
All paddles are serialized and designed to meet USP specifications for USP Apparatus 2 including an individual CoC as standard. Paddles are available in stainless steel, PTFE-coated, or PEEK in a range of standard, mega, or mini sizes.
: Each paddle includes an individual Certificate of Conformance as standard.
Specifications
- Description:Paddle, lower interchangeable Spin on/off, electropolished stainless steel, 1 L, 21", for 705-DS; 708-DS; 709-DS; VK7000
- Certification:Yes