Orders LinkContactUsLinkComponent
Klinger Light Box Multi Pack, 3 Colors
Klinger Light Box Multi Pack, 3 Colors
Klinger LED light box, red
Catalog # 470336-002
Supplier:   Klinger
Supplier #:  LTB-RD-KL
Klinger Light Box Multi Pack, 3 Colors
Catalog # 470336-002
Supplier:  Klinger
Klinger Light Box Multi Pack, 3 Colors
Klinger Light Box Multi Pack, 3 Colors
Klinger LED light box, red
Price
$39.19
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Beautiful, bright and captivating classroom safe LED's in primary colors of light (red, green and blue) for color mixing, refraction and reflection.
  • Bring in additive color with one fun and inexpensive set of light box LED's
  • With the light boxes attached to whiteboard it doubles as a ray box
  • Research, examine and detect all the properties of light with one uncomplicated tool
  • There is no need for expensive pieces of science apparatus to make the science of light and optics fun and available for ages 7 to 18 and higher
  • A hands-on science tool for the classroom
  • Light box requires 2 AA batteries, included
The primary colors of light - red, green and blue are not like the primary colors of pigments - when you mix all three primary colors of light, you get white light. All of the white light we live in every day is made up of many wavelengths of light, but our eyes are only able to distinguish red, green, and blue.
These light boxes project a bright and easy to see beam in each of the primary colors of light (red, green, and blue), perfect for examining how light ricochets (reflects) and twists (refracts). Students simply and naturally uncover the law of reflection or measure the angle of refraction.
These sturdy light boxes that are designed just for the classroom LED's are perfect for mixing up the secondary colors of light yellow, magenta and cyan as well as for mixing a pure white light.
Specifications
  • Description:
    Klinger LED light box, red