Orders LinkContactUsLinkComponent
Klinger Laser Boxes
Klinger Laser Boxes
Klinger laser box, green, single
Catalog # 470335-996
Supplier:   Klinger
Supplier #:  LB-GR-KL
Klinger Laser Boxes
Catalog # 470335-996
Supplier:  Klinger
Klinger Laser Boxes
Klinger laser box, green, single
Price
$116.09
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Perform many different lessons around reflection, refraction, diffraction, and interference.
  • Class 3R lasers
  • Line projection lens installed
  • Wavelengths: 635 nm (red), 532 nm (green), and 450 nm (blue)
  • 2× AA batteries included
Klinger lasers are classroom safe and are designed for teaching and learning about light, lasers, and optics. These lasers can be used just like a laser pointer or on a white board with their powerful magnets to keep them in place. Place multiple lasers together side by side to create a Ray Box for even more versatility and flexibility.
Specifications
  • Description:
    Klinger laser box, green, single