Orders LinkContactUsLinkComponent
Orum International Remote Head Air Sampling Systems
Orum International Remote Head Air Sampling Systems
Remote head air sampling system for contact plates
Catalog # 76509-276
Supplier:   Hardy Diagnostics
Supplier #:  BAS191
Orum International Remote Head Air Sampling Systems
Catalog # 76509-276
Supplier:  Hardy Diagnostics
Orum International Remote Head Air Sampling Systems
Orum International Remote Head Air Sampling Systems
Remote head air sampling system for contact plates
Price
$6,580.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Remote Head System for contact plates or Petri plates with tri-clamp connection. Completely fabricated in AISI 316 stainless steel.
  • Reduced risk of contamination associated with operator activity
  • Aspirating chamber can be manipulated in multiple directions, according to the unidirectional airflow
  • TRI-clamp device simplifies all the operations of cleaning/sterilization
  • Remote Head System is fully sterilizable
Microbial sampling is programmed through the control unit (Trio.Bas™, AIRBIO, or MULTIFLEX not included) which is generally located outside the isolator cabinet, RABS, or inside/outside the cleanroom. The operator must insert the plate with the culture media into the separated aspiration chamber.
Systems include stainless steel aspirating head (contact plate or Petri plate format), and one tube clamp end. All components are sold separately to allow for customization.
Specifications
  • Description:
    Remote head air sampling system for contact plates