Order Entry
United States
Orders LinkContactUsLinkComponent
 

FectoCHO® Expression System

FectoCHO™ CD Expression Medium, 6×1 L (Expression Medium), —
Catalog # 76234-743
 :  101000003
FectoCHO® Expression System
Catalog # 76234-743
CAS Number:  
FectoCHO® Expression System
FectoCHO® Expression System

FectoCHO™ CD Expression Medium, 6×1 L (Expression Medium), —

CAS Number:  
Price
$1,115.26
Log in for availability
Pack of 6
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

FectoCHO® Expression system consists in the synergistic association of a novel CHO chemically defined medium, FectoCHO® CD Expression medium and a powerful transfection reagent, FectoPRO®.

  • Outstanding protein production yields in various CHO cells
  • Fast and effortless CHO cell adaptation
  • Chemically defined kit components
  • Excellent lot to lot reproducibility
  • Cost-effective solution

FectoCHO® CD Expression medium allows easy cultivation of various strains of CHO cells such as CHO-K1, CHO-S and ExpiCHO®-S , without the need of an extensive sequential adaptation. FectoPRO®-mediated transient transfection in this chemically defined medium leads to the highest protein and antibody yields ever reached using Transient Gene Expression, up to 4 times higher than with the competitor's systems.

Certifications: Since 2002, Polyplus-transfection® is ISO 9001 certified and since then this certification has been successfully renewed every three years.

Ordering information: FectoPRO® transfection reagent, FectoPRO® Booster and FectoCHO® CD Expression Medium can be purchased separately.

Specifications

Specifications

  • Description:
    FectoCHO™ CD Expression Medium
  • Size:
    6×1 L (Expression Medium)
  • Buffer volume:

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.