Order Entry
United States
Orders LinkContactUsLinkComponent
Splitless Liners for GC

Splitless Liners for GC

Inlet liner, splitless, single taper, glass wool, deactivated, 6.47 mm, —, 900 µl, 4×78.5 mm
Catalog # AG5062-3587
Supplier #:  5062-3587
Splitless Liners for GC
Catalog # AG5062-3587
Splitless Liners for GC
Splitless Liners for GC

Inlet liner, splitless, single taper, glass wool, deactivated, 6.47 mm, —, 900 µl, 4×78.5 mm

Price
$52.70
Log in for availability
Each
Product Details & Documents

We offer a complete line of liners for your Agilent GC splitless inlet, with exclusive deactivation.

  • Feature Agilent’s exclusive deactivation
  • Designed and engineered to meet strict specifications for dimensional accuracy and inertness
  • Outer diameter of splitless liners is ground and polished to exacting dimensional tolerances, ensuring a perfect fit into our inlets and optimal splitless injection
  • We recommend trying a liner without glass wool first
  • Use glass wool as a second choice

Choose a liner without glass wool if your sample contains nonvolatiles or analytes within a wide boiling range. For unknown samples, use a liner without glass wool to avoid loss of active or labile compounds.

Agilent offers a complete portfolio of liners for your Agilent GC splitless inlet. Our exclusive deactivation is important because of the longer sample/liner contact in splitless mode.

Specifications
  • Ext.Ø:
    6.47 mm
  • Ø int.:
  • Volume:
    900 µl
  • Description:
    Inlet liner, splitless, single taper, glass wool, deactivated
  • I.D.×L:
    4×78.5 mm