Pioloform TEM Grids, Electron Microscopy Sciences
Product Details & Documents
Coated grids made with Pioloform, which offers the advantage of higher thermal stability.
- Approximately 5 to 6 nm
- All film laid upon the shiny side of the grid
Pioloform TEM grids feature a thin film of pioloform deposited on one side of the grid, no carbon layer, and less elecron scattreing. They have the advantage of higher thermal stability, as well as mechanical stability, over Formvar.
Specifications
- Description:Pioloform TEM grid, 300 mesh
- Material:Nickel