Orders LinkContactUsLinkComponent
Au-Flat Grids, Electron Microscopy Sciences
Au-flat cryo-em grid, 300 µm mesh size, Gold, 1.2 µm (1.3 µm spacing)
  76499-690
 :  AUFT313-50
Au-Flat Grids, Electron Microscopy Sciences
  76499-690
Au-Flat Grids, Electron Microscopy Sciences
Au-Flat Grids, Electron Microscopy Sciences
Au-flat cryo-em grid, 300 µm mesh size, Gold, 1.2 µm (1.3 µm spacing)
$1,683.40
Pack of 50
Au-Flat, ultrastable Cryo-EM sample support with a 45 nm holey Gold alloy film on 3 mm gold mesh grid.
  • Holey Au/Pd film on a gold mesh grid
  • Stronger than carbon films
  • More capable of surviving the Cryo-EM workflow
The Au-Flat Cryo-EM grid significantly reduces beam-induced motion during imaging compared to carbon films, improving both image quality and resolution.
  • Description:
    Au-flat cryo-em grid, 300 µm mesh size
  • Material:
    Gold
  • Hole size:
    1.2 µm (1.3 µm spacing)