Orders LinkContactUsLinkComponent
Au-Flat Grids, Electron Microscopy Sciences
Au-Flat Grids, Electron Microscopy Sciences
Au-flat cryo-em grid, 300 µm mesh size, Gold, 1.2 µm (1.3 µm spacing)
Catalog # 76499-688
Supplier #:  AUFT313-05
Au-Flat Grids, Electron Microscopy Sciences
Catalog # 76499-688
Au-Flat Grids, Electron Microscopy Sciences
Au-Flat Grids, Electron Microscopy Sciences
Au-flat cryo-em grid, 300 µm mesh size, Gold, 1.2 µm (1.3 µm spacing)
Price
$228.36
Log in for availability
Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Au-Flat, ultrastable Cryo-EM sample support with a 45 nm holey Gold alloy film on 3 mm gold mesh grid.
  • Holey Au/Pd film on a gold mesh grid
  • Stronger than carbon films
  • More capable of surviving the Cryo-EM workflow
The Au-Flat Cryo-EM grid significantly reduces beam-induced motion during imaging compared to carbon films, improving both image quality and resolution.
Specifications
  • Description:
    Au-flat cryo-em grid, 300 µm mesh size
  • Material:
    Gold
  • Hole size:
    1.2 µm (1.3 µm spacing)