Au-Flat Grids, Electron Microscopy Sciences
Product Details & Documents
Au-Flat, ultrastable Cryo-EM sample support with a 45 nm holey Gold alloy film on 3 mm gold mesh grid.
- Holey Au/Pd film on a gold mesh grid
- Stronger than carbon films
- More capable of surviving the Cryo-EM workflow
The Au-Flat Cryo-EM grid significantly reduces beam-induced motion during imaging compared to carbon films, improving both image quality and resolution.
Specifications
- Description:Au-flat cryo-em grid, 200 µm mesh size
- Material:Gold
- Hole size:2 µm (2 µm spacing)