Ice-Pak™ Foam Bricks, Cryopak Verification Technologies
Ice-Pak™ Foam bricks, 5.5×8.25×1.5" (×2)
Catalog # 76499-192
Supplier: Cryopak Verification Technologies
: FBRK00019
Ice-Pak™ Foam Bricks, Cryopak Verification Technologies
Catalog # 76499-192
Supplier: Cryopak Verification Technologies
Jump to:
Product Details & Documents
Vacuum-sealed in durable film, these bricks are ideal for defined and precise packaging configurations.
- Durable materials
- Reusable
- Consistent structure
Ice-Pak™ foam bricks have a freezing/melting point of 0 °C and offer superior thermal performance when transporting temperature-sensitive products. They feature a consistent, uniform structure that is maintained even after freezing.
Specifications
- Description:Ice-Pak™ Foam bricks
- L×W×D:5.5×8.25×1.5" (×2)