FLASK T-175 PS TISSUE CULTURE CS100
Catalog # MSPP-TC30-130A
Supplier: STELLAR SCIENTIFIC MS
FLASK T-175 PS TISSUE CULTURE CS100
Catalog # MSPP-TC30-130A
Supplier: STELLAR SCIENTIFIC MS
Supplier Number: TC30-130
Frequently Bought Together
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Xylene (mixture of isomers) for histology
Supplier: Honeywell Research Chemicals