Paraffin Tapper
Designed to flatten tissue during the histology embedding process.
Stainless steel construction for durability and corrosion resistance
- Ergonomic handle for comfortable, precise operation
- Easy to clean and maintain for long-term use
- Essential tool for pathology and histology workflows
Constructed from durable, laboratory-grade stainless steel, the Mopec Paraffin Tapper offers long-lasting performance and resistance to corrosion. Its ergonomic handle ensures a comfortable, controlled grip, reducing hand fatigue during repetitive use. The balanced design provides excellent control, making it ideal for use in histology, pathology, and research laboratories.
- Description:Stainless steel paraffin tapper, 29×23×1.5 mm