Order Entry
United States
Orders LinkContactUsLinkComponent
Economy Platform Scale, Mortech®

Economy Platform Scale, Mortech®

Economy platform scale, 5000 g, 1 g
Catalog # 76498-274
Supplier:   Mopec
Supplier #:  BB062
Economy Platform Scale, Mortech®
Catalog # 76498-274
Supplier:  Mopec
Economy Platform Scale, Mortech®
Economy Platform Scale, Mortech®

Economy platform scale, 5000 g, 1 g

Price
$483.20
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Economy platform scales are available in 2000 or 5000 g weighing capacities.

  • 6" (15.2 cm) diameter platform
  • Gram/lbs./oz. weighing modes, (1 g/0.05 oz. readability)
  • Easily portable, tare function
  • Battery or AC power options
  • One year warranty
Specifications
  • Description:
    Economy platform scale
  • Readability:
    1 g
  • Weighing capacity:
    5000 g

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.