Orders LinkContactUsLinkComponent
Dissection Needles
Dissection Needles
Needle for needle holder, -, -, -
Catalog # 470334-822
Supplier:   AFTAB AHMED SE
Supplier #:  14-0633
Dissection Needles
Catalog # 470334-822
Supplier:  AFTAB AHMED SE
Dissection Needles
Dissection Needles
Needle for needle holder, -, -, -
Price
$3.10
Log in for availability
Each Box of 100
Product Details & Documents
Sharp point needles.
  • Fine point teasing needles
  • Straight or angled tips
Very sharp points on these needles make tissue separation fast and easy. Used mostly to break apart muscle tissues.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Description:
    Needle for needle holder
  • Length:
  • Material:
    -
  • Tip style:
Recommendations