TLRB AGONIST SSPOLYU/LYOVEC 4X25UG
Catalog # MSPP-TLRL-LPU
Supplier: Invivogen
TLRB AGONIST SSPOLYU/LYOVEC 4X25UG
Catalog # MSPP-TLRL-LPU
Supplier: Invivogen
Supplier Number: TLRL-LPU
Frequently Bought Together
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International