Trajectory Kit
Protrusion rods, small (0.22 cal), —
Catalog # 470334-330
Supplier: FORENSICS SOURCE
Supplier #: 1007801
Jump to:
Product Details & Documents
Includes Everything Needed to Measure Trajectories at Crime Scenes.
- Reuseable Materials
- Math and Physics Intensely Used
Accurately measure angles and slopes every time with professional forensics equipment, including four 18L×¹/₄" diameter, yellow aluminum protrusion rods with connectors, to provide sufficient length for easy trajectory measurements; two 1" long and two 2" long spacer cones for placement and centering of protrusion rods in various sized holes; and a level and angle finder that is easily mounted to protrusion rods or the included aluminum tripod mount with universal thread. Available to U.S. customers only, similar product will ship elsewhere.
Specifications
- Description:Protrusion rods, small (0.22 cal)
- No. of lab groups:—