ASI Color Staph Latex Test Kits, Arlington Scientific
Catalog # 76474-210
Supplier: Arlington Scientific
CAS Number:
Specifications
- Description:50-test kit
- Cat. No.:76474-210
Specifications
About this item
The ASI Color Staph is a rapid, color enhanced latex agglutination test to detect clumping factor and/or Protein-A, two characteristics associated with Staphylococcus aureus colonies obtained from culture.
- Simple to interpret
- 10 to 30 second slide test
- Detects Methicillin Resistant S. aureus (MRSA)
- Liquid controls included
- Longest shelf life of up to 24 months
- Made in USA
ASI Color Staph latex reagent will react with either or both of these characteristics. The aggregation of the smooth latex suspension represents a positive reaction which is visible to the unaided eye producing a red agglutination in a 'blue' background.
Certifications: CPT Code 87449
510(k)-K830783
510(k)-K830783
Frequently Bought Together