BUFFER RIPA LYSIS STORE AT 4 DEG C 10 ML
Catalog # MSPP-040-483
Supplier: PROTEIN SIMPLE MS
BUFFER RIPA LYSIS STORE AT 4 DEG C 10 ML
Catalog # MSPP-040-483
Supplier: PROTEIN SIMPLE MS
Supplier Number: 040-483
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Protease Inhibitor Cocktails
Supplier: VWR
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
ANTIBODY CRISPR CAS9 POLYCLONAL 100UL
Supplier: Diagenode
Cholera Toxin, <i>Vibrio cholerae</i>, Type Inaba 569B, Azide Free, MilliporeSigma
Supplier: MilliporeSigma
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
BUFFER SAMPLE DILUENT BICINE/CHAPS LYSIS
Supplier: PROTEIN SIMPLE MS
Halt™ Protease and Phosphatase Inhibitor Cocktails, Thermo Scientific
Supplier: Invitrogen
TRIS WASH BUFFER (10X) 1000ML
Supplier: Meso Scale Diagnostics
ANTIBODY ANTI-RABBIT SECONDARY
Supplier: PROTEIN SIMPLE MS
ASSAY RPLEX HUMAN KALLIKREIN K151ACJR-2
Supplier: Meso Scale Diagnostics
Q5® Hot Start High-Fidelity 2X Master Mix
Supplier: New England Biolabs (NEB)
KIT ISCRPT CDNA SYNTH 100
Supplier: Bio-Rad
Halt™ Protease Inhibitor Cocktail (100X), Thermo Scientific
Supplier: Invitrogen