QUICK-RNA VIRAL 96 KIT DX 2 X 96 PREPS
Catalog # 76469-090
Supplier: Zymo Research
QUICK-RNA VIRAL 96 KIT DX 2 X 96 PREPS
Catalog # 76469-090
Supplier: Zymo Research
Supplier Number: R1040-E
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Cell Culture Storage Bottles, Polystyrene, Corning
Supplier: Corning
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
VWR® Razor Blades
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Reagent Reservoirs
Supplier: VWR International