RIPA LYSIS BUFFER 100ML
Catalog # 76465-956
Supplier: Boster Biological Technology
RIPA LYSIS BUFFER 100ML
Catalog # 76465-956
Supplier: Boster Biological Technology
Supplier Number: AR0105-100
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Multiwell Cell Culture Plates, Corning
Supplier: BD
Tissue-Tek® Accu-Edge® Disposable Microtome Blades, Sakura® Finetek
Supplier: Sakura Finetek
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
Laemmli SDS sample buffer, reducing (4X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
Scissors, Sharp, Fine Science Tool
Supplier: 0000013460