Ward's® Pure Preserved Squids
Pure preserved squid, 12"+, Single injected, Pack of 1
Catalog # 470330-502
Supplier: Avantor
Jump to:
Product Details & Documents
Ward's® PURE Preserved Specimens - the best dissection experience from start to finish.
- Safest specimens
- Low formalin odor
- Best classroom experience
Ward's® PURE preserved specimens offer the best classroom experience because PURE specimens are safe. Students stay focused on learning and not on smells. Experience quality with true-to-life preservation, combined with artfully illustrated dissection guides, free with every purchase, to maximize value, lesson time and student engagement.
We guarantee you’ve never experienced dissection like this before. Go Pure.
Loligo or ommastrephes. The squid has the most highly developed nervous system of all the invertebrates. Dissection shows developed eyes and nervous system.
Specifications
- Description:Pure preserved squid
- Size:12"+
- Injection type:Single injected
- Packaging:Pack of 1