FLOW STAINING BUFFER 1X 500ML
Catalog # 76448-862
Supplier: Tonbo Biosciences
FLOW STAINING BUFFER 1X 500ML
Catalog # 76448-862
Supplier: Tonbo Biosciences
Supplier Number: TNB-4222-L500
Frequently Bought Together
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Disposable Serological Pipettes
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Vi-CELL™ MetaFLEX Sensor Cassette, Beckman Coulter
Supplier: Beckman Coulter
SpeI-HF®, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs (NEB)
ASSAY DNA FLUOROCHROME STAINING 2ML
Supplier: BIONIQUE TESTING LABORATORY MS
SOC Outgrowth Medium, New England Biolabs
Supplier: New England Biolabs (NEB)
SphI-HF®, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs (NEB)