Orders LinkContactUsLinkComponent
Tomography Grids, Electron Microscopy Sciences
Tomography Grids, Electron Microscopy Sciences
Lacey carbon
Catalog # 76439-536
Supplier #:  74357-01
Tomography Grids, Electron Microscopy Sciences
Catalog # 76439-536
Tomography Grids, Electron Microscopy Sciences
Tomography Grids, Electron Microscopy Sciences
Lacey carbon
Price
$525.88
Log in for availability
Pack of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This 1.5 mm square, 300 mesh grid is made from copper and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90°.
Specifications
  • Type:
    Lacey carbon