Orders LinkContactUsLinkComponent
Picking Trays, Electron Microscopy Sciences
Picking Trays, Electron Microscopy Sciences
Picking tray, Aluminum, tan, 96×34 mm (3.78×1.34")
Catalog # 76439-518
Supplier #:  71171-03
Picking Trays, Electron Microscopy Sciences
Catalog # 76439-518
Picking Trays, Electron Microscopy Sciences
Picking Trays, Electron Microscopy Sciences
Picking tray, Aluminum, tan, 96×34 mm (3.78×1.34")
Price
$64.26
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Trays perfect for placement of grids and storage of small components. The trays feature 42 cells with a lip differentiating each cell. The cells measure 11×11 mm (⁷/₁₆×⁷/₁₆").
These unique trays are available in either black rigid ABS plastic or aluminum in black or tan.
Specifications
  • Description:
    Picking tray, Aluminum, tan
  • Dimensions:
    96×34 mm (3.78×1.34")