BUFFER 10X R STREPTACTIN WITH HABA 100ML
Catalog # MSPP-21002-100
Supplier: IBA MS
BUFFER 10X R STREPTACTIN WITH HABA 100ML
Catalog # MSPP-21002-100
Supplier: IBA MS
Supplier Number: 2-1002-100
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Culture Tubes, Plastic, with Dual-Position Caps
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Strep-Tactin® XT Sepharose Chromatography Resins, Cytiva
Supplier: Cytiva
Pierce™ BCA® Protein Assay Kits and Reagents, Thermo Scientific
Supplier: Invitrogen