PTS CARTRIDGES PK25
Catalog # MSPP-PTS5501F
Supplier: CHARLES RIVER ENDOSAFE MS
PTS CARTRIDGES PK25
Catalog # MSPP-PTS5501F
Supplier: CHARLES RIVER ENDOSAFE MS
Supplier Number: PTS5501F
Frequently Bought Together
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polypropylene, Avantor®
Supplier: VWR International
VWR® Traceable® Giant-Digit™ Countdown Timer
Supplier: VWR International
VWR® 80-Well Microtube Racks
Supplier: VWR International
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
IRRIGATED WATER STERILE USP 1L
Supplier: B Braun
Potassium carbonate ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
Heavy Duty Aluminum Foil, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Poloxamer 188
Supplier: Thermo Scientific Chemicals
Thymidine (2'-deoxythymidine) ≥99%
Supplier: BeanTown Chemical
StrepTrap XT Prepacked Chromatography Columns, Cytiva
Supplier: Cytiva
Mouse Recombinant Klotho (from CHO Cells)
Supplier: R&D Systems
Bioquell Hydrogen Peroxide Sterilant Case
Supplier: Eco Lab
AcquaStain Protein Gel Stain, Bulldog Bio
Supplier: Bulldog Bio
Water nuclease-free
Supplier: Promega Corporation
Endosafe® PTS™ Endotoxin Tests, Charles River
Supplier: CHARLES RIVER ENDOSAFE MS