RNA CLEAN/CONCENTRATOR MAGBEAD 96PREPS
Catalog # 76421-018
Supplier: Zymo Research
RNA CLEAN/CONCENTRATOR MAGBEAD 96PREPS
Catalog # 76421-018
Supplier: Zymo Research
Supplier Number: R1081
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
VWR® Disposable Serological Pipettes
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
sparQ PureMag Beads, Quantabio
Supplier: Quantabio
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
Microflex® Onyx® N64 Nitrile Disposable Gloves
Supplier: Ansell Healthcare
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
Direct-zol™ RNA Kits, Zymo Research
Supplier: Zymo Research