ANTI IBA1 RABBIT SPICA DYETM 568-C 100UL
Catalog # MSPP-015-28011
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
ANTI IBA1 RABBIT SPICA DYETM 568-C 100UL
Catalog # MSPP-015-28011
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
ANTIBODY IBA1 SPICA DYE568-CONJD 100UL
Supplier: FUJIFILM IRVINE SCIENTIFIC WAKO CHE
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
Beakers, Stainless Steel, Ace Glass Incorporated
Supplier: Ace Glass