Order Entry
United States
Orders LinkContactUsLinkComponent
D-(-)-Glutamic acid 98%

D-(-)-Glutamic acid 98%

Glass bottle, 5 g
  103848-728
 
Marketsource
D-(-)-Glutamic acid 98%
  103848-728
 :  
Marketsource
 :  6893-26-1
D-(-)-Glutamic acid 98%

Glass bottle, 5 g

 :  6893-26-1
 
 
 
D-GLUTAMIC ACID 25GM  TCG0057-025G

Product TCG0057-025G is similar to 103848-728 but manufacturer and specifications may vary.

N-ALPHA-BENZOYL-DL-ARGININE-4-NITR 1G  TS22774-0010

Product TS22774-0010 is similar to 103848-728 but manufacturer and specifications may vary.

D-GLUTAMIC ACID 99+% 5G  AAA14191-06

Product AAA14191-06 is similar to 103848-728 but manufacturer and specifications may vary.

 
   
 

Caution: for Research Use Only

 :  (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
  •  :6893-26-1
  •  :C₅H₉NO₄
  •  :MFCD00063112
  •  :147.13 g/mol
  •  :333.8 °C (760 mm Hg)
  •  :200…202 °C
  •  :155.7 °C
  •  :Ambient
 

 

  • Pack type:
    Glass bottle
  • Size:
    5 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.