Order Entry
United States
Orders LinkContactUsLinkComponent
 

3-(3-Fluorophenyl)propionic acid 98%

Glass bottle, 5 g
  103843-716
 
Marketsource
3-(3-Fluorophenyl)propionic acid 98%
  103843-716
 :  
Marketsource
 :  458-45-7
3-(3-Fluorophenyl)propionic acid 98%
3-(3-Fluorophenyl)propionic acid 98%

Glass bottle, 5 g

 :  458-45-7
 
 
 
3- 3-FLROPHENL PROPIONIC AC 5G  AAH27847-06

Product AAH27847-06 is similar to 103843-716 but may have a different manufacturer and/or packaging.

3-(3-FLUOROPHENYL)PROPIONIC ACID 98% 5G  77319-690

Product 77319-690 is similar to 103843-716 but may have a different manufacturer and/or packaging.

3-(3-FLUOROPHENYL)PROPIONIC 5G  TCF0779-5G

Product TCF0779-5G is similar to 103843-716 but may have a different manufacturer and/or packaging.

 
 
   
 

Caution: for Research Use Only

 :  3-(3-Fluorophenyl)propanoic acid
  •  :458-45-7
  •  :C₉H₉FO₂
  •  :MFCD01310821
  •  :168.17
  •  :270.6 °C (760 mm Hg)
  •  :43…47 °C
  •  :117.4 °C
  •  :Ambient
 

 

  • Pack type:
    Glass bottle
  • Size:
    5 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.