Order Entry
United States
Orders LinkContactUsLinkComponent
Lenalidomide 98%

Lenalidomide 98%

Glass bottle, 5 g
Catalog # 103837-452
Health
Marketsource
Lenalidomide 98%
Catalog # 103837-452
Supplier:  
Marketsource
CAS Number:  191732-72-6
Lenalidomide 98%

Glass bottle, 5 g

CAS Number:  191732-72-6
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
LENALIDOMIDE 97+% CRYSTAL 5G Catalog #76840-696

Product 76840-696 is similar to 103837-452 but manufacturer and specifications may vary.

(3S)-3-(4-AMINO-1-OXO-1 5G Catalog #102551-654

Product 102551-654 is similar to 103837-452 but manufacturer and specifications may vary.

LENALIDOMIDE 97% 25G Catalog #103890-368

Product 103890-368 is similar to 103837-452 but manufacturer and specifications may vary.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Caution: for Research Use Only

Synonyms:  3-(4-Amino-1-oxo-1,3-dihydro-2H-isoindol-2-yl)piperidine-2,6-dione, (R,S)-Lenalidomide, 3-(4-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione
  • CAS Number:191732-72-6
  • Formula:C₁₃H₁₃N₃O₃
  • MDL Number:MFCD07772307
  • Molecular Weight:259.27 g/mol
  • Boiling Pt:614 °C (760 mm Hg)
  • Melting Pt:269…271 °C
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.