Order Entry
United States
Orders LinkContactUsLinkComponent
D-(+)-Mannose 95%

D-(+)-Mannose 95%

Glass bottle, 5 g
  103835-496
 
Marketsource
D-(+)-Mannose 95%
  103835-496
 :  
Marketsource
 :  3458-28-4
D-(+)-Mannose 95%

Glass bottle, 5 g

 :  3458-28-4
 
 
 
D-(+)-MANNOSE 99% 25G  AAA10842-14

Product AAA10842-14 is a functional alternative to 103835-496 but Packaging, Quantity, Manufacturer and/or specifications may vary.

D-(+)-MANNOSE 25GM  TCM0045-025G

Product TCM0045-025G is a functional alternative to 103835-496 but Packaging, Quantity, Manufacturer and/or specifications may vary.

 
   
 

Caution: for Research Use Only

 :  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  •  :3458-28-4
  •  :C₆H₁₂O₆
  •  :MFCD00064122
  •  :180.16 g/mol
  •  :410.8 °C (760 mm Hg)
  •  :133…140 °C
  •  :202.2 °C
  •  :Ambient
 

 

  • Pack type:
    Glass bottle
  • Size:
    5 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.