RNA TOTAL MONKEY LIVER CYNOMOLGUS 0.1MG
Catalog # MSPP-KR-314
Supplier: ZYAGEN MS
RNA TOTAL MONKEY LIVER CYNOMOLGUS 0.1MG
Catalog # MSPP-KR-314
Supplier: ZYAGEN MS
Supplier Number: KR-314
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
TapeStation Systems Parts and Accessories
Supplier: Agilent Technologies
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
TapeStation Parts and Accessories for 2200
Supplier: Agilent Technologies