Order Entry
United States
Orders LinkContactUsLinkComponent
3-(3-Fluorophenyl)propionic acid 98%

3-(3-Fluorophenyl)propionic acid 98%

Glass bottle, 1 g
Catalog # 103805-478
Irritant
Marketsource
3-(3-Fluorophenyl)propionic acid 98%
Catalog # 103805-478
Supplier:  
Marketsource
CAS Number:  458-45-7
3-(3-Fluorophenyl)propionic acid 98%
3-(3-Fluorophenyl)propionic acid 98%

Glass bottle, 1 g

CAS Number:  458-45-7
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
3-(3-FLUOROPHENYL)PROPIONIC A Catalog #101790-116

Product 101790-116 is similar to 103805-478 but manufacturer and specifications may vary.

3-(3-FLUOROPHENYL)PROPIONIC ACID 98% 1G Catalog #77319-686

Product 77319-686 is similar to 103805-478 but manufacturer and specifications may vary.

3-(3-FLUOROPHENYL)PROPIONIC 1G Catalog #TCF0779-1G

Product TCF0779-1G is similar to 103805-478 but manufacturer and specifications may vary.

See more alternatives
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

 : for Research Use Only

Synonyms:  3-(3-Fluorophenyl)propanoic acid
  • CAS Number:458-45-7
  • Formula:C₉H₉FO₂
  • MDL Number:MFCD01310821
  • Molecular Weight:168.17
  • Boiling Pt:270.6 °C (760 mm Hg)
  • Melting Pt:43…47 °C
  • Flash Pt:117.4 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.