Order Entry
United States
Orders LinkContactUsLinkComponent
 

4'-Bromo-2'-hydroxyacetophenone 98%

Glass bottle, 1 g
Catalog # 103803-946
Irritant
Marketsource
4'-Bromo-2'-hydroxyacetophenone 98%
Catalog # 103803-946
Supplier:  
Marketsource
CAS Number:  30186-18-6
4'-Bromo-2'-hydroxyacetophenone 98%

Glass bottle, 1 g

CAS Number:  30186-18-6
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
4'-BRMO-2'-HYDRXYACETOPHEN1 1G Catalog #AAH61137-03

 

1-(4-BROMO-2-HYDROXYPHEN 1G Catalog #CH24972-1G

 

See more alternatives
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

 : for Research Use Only

Synonyms:  1-(4-Bromo-2-hydroxyphenyl)ethanone, 1-(4-Bromo-2-hydroxyphenyl)ethan-1-one
  • CAS Number:30186-18-6
  • Formula:C₈H₇BrO₂
  • MDL Number:MFCD03428533
  • Molecular Weight:215.05
  • Boiling Pt:287.7 °C (760 mm Hg)
  • Melting Pt:38…41 °C
  • Flash Pt:127.8 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.