Order Entry
United States
Orders LinkContactUsLinkComponent
D-(-)-Glutamic acid 98%

D-(-)-Glutamic acid 98%

Glass bottle, 1000 g
Catalog # 103809-456
Irritant
Marketsource
D-(-)-Glutamic acid 98%
Catalog # 103809-456
Supplier:  
Marketsource
CAS Number:  6893-26-1
D-(-)-Glutamic acid 98%

Glass bottle, 1000 g

CAS Number:  6893-26-1
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
D-GLUTAMIC ACID MF=C5H9 500G Catalog #102833-828

Product 102833-828 is a functional alternative to 103809-456 but Packaging, Quantity, Manufacturer and/or specifications may vary.

D-GLUTAMIC ACID Catalog #TCG0057-250G

Product TCG0057-250G is a functional alternative to 103809-456 but Packaging, Quantity, Manufacturer and/or specifications may vary.

D-GLUTAMIC ACID 99+% 100G Catalog #AAAA14191-22

Product AAAA14191-22 is a functional alternative to 103809-456 but Packaging, Quantity, Manufacturer and/or specifications may vary.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Caution: for Research Use Only

Synonyms:  (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
  • CAS Number:6893-26-1
  • Formula:C₅H₉NO₄
  • MDL Number:MFCD00063112
  • Molecular Weight:147.13 g/mol
  • Boiling Pt:333.8 °C (760 mm Hg)
  • Melting Pt:200…202 °C
  • Flash Pt:155.7 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1000 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.