RackScan® 2D Barcode Scanner with RackScan Analyzer Software, Eppendorf®
2D barcode scanner, bottom reading
Catalog # 76404-530
Supplier: Eppendorf
Supplier #: 6002000103
RackScan® 2D Barcode Scanner with RackScan Analyzer Software, Eppendorf®
Catalog # 76404-530
Supplier: Eppendorf
Jump to:
Product Details & Documents
The more samples you have, the less you want to scan them manually. The Eppendorf RackScan system enables scanning of up to 96 cryo storage Vials at one time.
- Camera-based instrument for rapid imaging and decoding
- ECC 200 error correction function enables safe code reading even when codes are up to 30% damaged
- Simple and easy-to-use plug-in system
- Small footprint, ideal for benchtop (and integration)
To scan barcodes located on the side of vials, the accessory Rackscan side scanner is available as an optional add-on. RackScan analyzer software reads the barcodes of your vials to provide the information to downstream applications, either via manual import into local databases or automatically via the Eppendorf eLABInventory software (sold separately).
Specifications
- Description:2D barcode scanner, bottom reading